Learn More
Medchemexpress LLC Clonidine hydrochloride | 4205-91-8 | MFCD00036705 | 99.9% | 500 MG
SDP
Catalog No. 5000407872
Encompass_Preferred
Clonidine hydrochloride is an agonist of α2-adrenoceptor and a potent antihypertensive agent. It is intended for research use only.
  • Agonist of α2-adrenoceptor
  • Potent antihypertensive agent
  • Induces CGRP mRNA expression
  • Increases NO level in endothelial cells
  • Decreases body temperature in rats
  • Suppresses dopamine efflux in the prefrontal cortex
Catalog No. 50-004-07872 Supplier Medchemexpress LLC Supplier No. HYB0409A500MG
Clonidine hydrochloride is an agonist of α2-adrenoceptor and a potent antihypertensive agent. It is intended for research use only.
  • Agonist of α2-adrenoceptor
  • Potent antihypertensive agent
  • Induces CGRP mRNA expression
  • Increases NO level in endothelial cells
  • Decreases body temperature in rats
  • Suppresses dopamine efflux in the prefrontal cortex