Learn More
TARGETMOL CHEMICALS INC Clonidine hydrochloride 500MG
SDP
Catalog No. 502879515
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Clonidine hydrochloride (Catapres) is a centrally active alpha-adrenergic agonist used predominantly as an antihypertensive agent. Purity 99.8%
Catalog No. 50-287-9515 Supplier TARGETMOL CHEMICALS INC Supplier No. T1247500MG
Also available in 1 mL, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. Clonidine hydrochloride (Catapres) is a centrally active alpha-adrenergic agonist used predominantly as an antihypertensive agent. Purity 99.8%