Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clonidine Hydrochloride, USP, 98.5-101%, Spectrum™ Chemical
SDP

Catalog No. 18601759 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-759 1 kg Amber Glass Bottle
18-601-755 1 g Amber Glass Bottle
18-601-756 5 g Amber Glass Bottle
18-601-757 25 g Amber Glass Bottle
18-601-758 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-601-759 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. CL1181KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

CL118, 4205-91-8, C9H9Cl2N3·HCl

Spectrum™ Chemical Clonidine Hydrochloride, USP is used alone or with other medications to treat high blood pressure. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 4205-91-8
Molecular Formula C9H10Cl3N3
Molecular Weight (g/mol) 266.55
InChI Key ZNIFSRGNXRYGHF-UHFFFAOYSA-N
IUPAC Name hydrogen N-(2,6-dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine chloride
SMILES [H+].[Cl-].ClC1=CC=CC(Cl)=C1NC1=NCCN1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 1 kg
UN Number UN2811
Percent Purity 98.5-101%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Clonidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.