Learn More
Boca Scientific Inc BriClone Hybridoma Cloning Additive, 100ml
Catalog No. 501685623
Encompass
BriClone is an additive for the cloning medium used in the post-fusion stages of hybridoma cell cloning. BriClone has been specifically developed by the NICB's dedicated team to overcome common problems in the post-fusion period such as batch-to-batch variation of feeder cells, and competition between feeder cells and freshly fused hybridomas for nutrients in the culture medium. To use BriClone, add to hybridoma culture media as a 5% (v/v) supplement. BriClone is designed for use in the post-fusion stages of hybridoma production and when cloning hybridoma cell lines under conditions of limiting dilution. BriClone, when added to culture medium eliminates the need for feeder cells in these types of cultures.
Catalog No. 50-168-5623 Supplier Boca Scientific Inc Supplier No. BRI100
BriClone is an additive for the cloning medium used in the post-fusion stages of hybridoma cell cloning. BriClone has been specifically developed by the NICB's dedicated team to overcome common problems in the post-fusion period such as batch-to-batch variation of feeder cells, and competition between feeder cells and freshly fused hybridomas for nutrients in the culture medium. To use BriClone, add to hybridoma culture media as a 5% (v/v) supplement. BriClone is designed for use in the post-fusion stages of hybridoma production and when cloning hybridoma cell lines under conditions of limiting dilution. BriClone, when added to culture medium eliminates the need for feeder cells in these types of cultures.