Learn More
Bellco Glass, Inc. Cloning Cylinder,10x10mm
SDP
Catalog No. 501945421
Encompass_Preferred
Used to surround a single cell to create homogeneous culture of cells rather than mixed population. The flat end ensures tight sealing with surface of culture dish. MADE IN THE USA. Sizes: 10x10mm
Catalog No. 50-194-5421 Supplier Bellco Glass, Inc. Supplier No. 209001010
Used to surround a single cell to create homogeneous culture of cells rather than mixed population. The flat end ensures tight sealing with surface of culture dish. MADE IN THE USA. Sizes: 10x10mm