Learn More
Chemglass Life Sciences Cloning Cylinders, 10 x 10mm, 125pcs/Pack
SDP
Catalog No. 501532241
Encompass_Preferred
Cloning Cylinders, 10 x 10mm, 125pcs/Pack. Clone a group or single cell by surrounding them with these glass cylinders. Cylinders are sealed to the plate with sterile silicone high vacuum grease, such as Dow Corning 976v thereby forming an isolation well. Cylinders are autoclavable or can be dry heat sterilized. Supplied in packages of 125 pieces.
Catalog No. 50-153-2241 Supplier Chemglass Life Sciences Supplier No. CLS177703
Cloning Cylinders, 10 x 10mm, 125pcs/Pack. Clone a group or single cell by surrounding them with these glass cylinders. Cylinders are sealed to the plate with sterile silicone high vacuum grease, such as Dow Corning 976v thereby forming an isolation well. Cylinders are autoclavable or can be dry heat sterilized. Supplied in packages of 125 pieces.