Learn More
Bellco Glass, Inc. Cloning Cylinder,6x8mm
SDP
Catalog No. 501945419
Encompass_Preferred
Used to surround a single cell to create homogeneous culture of cells rather than mixed population. The flat end ensures tight sealing with surface of culture dish. MADE IN THE USA. Sizes: 6x8mm 125 per case
Catalog No. 50-194-5419 Supplier Bellco Glass, Inc. Supplier No. 209000608
Used to surround a single cell to create homogeneous culture of cells rather than mixed population. The flat end ensures tight sealing with surface of culture dish. MADE IN THE USA. Sizes: 6x8mm 125 per case