Learn More
Bellco Glass, Inc. Cloning Cylinder,8x8mm
SDP
Catalog No. 501945420
Encompass_Preferred
Used to surround a single cell to create homogeneous culture of cells rather than mixed population. The flat end ensures tight sealing with surface of culture dish. MADE IN THE USA. Sizes: 8x8mm
Catalog No. 50-194-5420 Supplier Bellco Glass, Inc. Supplier No. 209000808
Used to surround a single cell to create homogeneous culture of cells rather than mixed population. The flat end ensures tight sealing with surface of culture dish. MADE IN THE USA. Sizes: 8x8mm