Learn More
Research Products International Corp Polystyrene Cloning Cylinders, Sterile, 60 per Case
SDP
Catalog No. 50212731
Encompass
Cloning cylinders improve cell production by assuring homogenous cultures and eliminating mixed populations. Cell growth starts in a sterile petri dish and the cylinders are added to isolate specific colonies. Cells are easily removed by adding a small amount of Trypsin, then incubating for a short period. Polystyrene cylinders are supplied sterile.
Catalog No. 50-212-731 Supplier Research Products International Corp Supplier No. 437847
Cloning cylinders improve cell production by assuring homogenous cultures and eliminating mixed populations. Cell growth starts in a sterile petri dish and the cylinders are added to isolate specific colonies. Cells are easily removed by adding a small amount of Trypsin, then incubating for a short period. Polystyrene cylinders are supplied sterile.