Learn More
Takara Bio CLONING ENHANCER 100 RXNS 6396
SDP
Catalog No. NC9744799
Encompass
Clontech's Cloning enhancer offers convenient treatment of PCR products without purification. Learn more
Catalog No. NC9744799 Supplier Takara Bio Supplier No. 639615
Clontech's Cloning enhancer offers convenient treatment of PCR products without purification. Learn more