Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clonixin | 17737-65-4 | 98.0% | 500 MG
SDP

Catalog No. 5000232576
Encompass_Preferred

Clonixin (SCH-10304) is a non-steroidal anti-inflammatory research compound provided for laboratory use as a solid or in DMSO solution. It is intended for in vitro and preclinical studies and is not for human or clinical use.

  • CAS number 17737-65-4.
  • Purity ≥98.0% (HPLC).
  • Molecular formula C13H11ClN2O2; molecular weight 262.69 g/mol.
  • Available as a 500 mg solid and as a 10 mM solution in DMSO.
  • Includes data sheet, certificate of analysis, and safety data sheet.
  • Common applications: in vitro pharmacology and anti-inflammatory assays.

Catalog No. 50-002-32576 Supplier Medchemexpress LLC Supplier No. HY121235500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clonixin (SCH-10304) is a non-steroidal anti-inflammatory research compound provided for laboratory use as a solid or in DMSO solution. It is intended for in vitro and preclinical studies and is not for human or clinical use.

  • CAS number 17737-65-4.
  • Purity ≥98.0% (HPLC).
  • Molecular formula C13H11ClN2O2; molecular weight 262.69 g/mol.
  • Available as a 500 mg solid and as a 10 mM solution in DMSO.
  • Includes data sheet, certificate of analysis, and safety data sheet.
  • Common applications: in vitro pharmacology and anti-inflammatory assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.