Learn More
Enzo Life Sciences Clopamide (500mg). CAS: 636-54-4
SDP
Supplier:  Enzo Life Sciences BMLCM1120500
Encompass_Preferred
Clinically useful diuretic. Clopamide selectively inhibits the chloride the sodium chloride cotransporter. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (20 mg/ml) or ethanol (5 mg/ml warm). Long Term Storage: Ambient.
Catalog No. 50-201-0416