Learn More
TARGETMOL CHEMICALS INC CLOPAMIDE 50MG
SDP
Catalog No. 502579577
Encompass_Preferred
Also available in 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Clopamide (Brinaldix) is a piperidine and sulfamoylbenzamide-based diuretic with thiazide-like diuretic activity. purity: 98%
Catalog No. 50-257-9577 Supplier TARGETMOL CHEMICALS INC Supplier No. T608650MG
Also available in 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Clopamide (Brinaldix) is a piperidine and sulfamoylbenzamide-based diuretic with thiazide-like diuretic activity. purity: 98%