Learn More
Medchemexpress LLC HY-B2179 500mg Medchemexpress, Cloperastine fendizoate CAS:85187-37-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB2179500MG
Encompass_Preferred
Medchemexpress, HY-B2179 500mg Cloperastine fendizoate CAS:85187-37-7 Cloperastine fendizoate inhibits the hERG K+ currents in a concentration-dependent manner with an IC50 value of 27 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2732