Learn More
Apexbio Technology LLC Clopidogrel, 100mg. Cas: 120202-66-6 MFCD: MFCD00876395. P2Y12 receptor antagonist
SDP
Supplier:  Apexbio Technology LLC A5183100
Encompass_Preferred
Selective, high affinity P2Y12 receptor antagonist. Inhibits ADP-induced platelet aggregation and displays antithrombotic activity. Active enantiomer of ()-clopidogrel hydrochloride. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4948