Learn More
Medchemexpress LLC HY-15283 100mg Medchemexpress, Clopidogrel CAS:113665-84-2 Purity:>98%
SDP
Catalog No. 502030605
Encompass_Preferred
Medchemexpress, HY-15283 100mg Clopidogrel CAS:113665-84-2 Clopidogrel is an orally active platelet inhibitor that targets P2Y12 receptor. Clopidogrel is used to inhibit blood clots in coronary artery disease, peripheral vascular disease, and cerebrovascular disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0605 Supplier Medchemexpress LLC Supplier No. HY15283100MG
Medchemexpress, HY-15283 100mg Clopidogrel CAS:113665-84-2 Clopidogrel is an orally active platelet inhibitor that targets P2Y12 receptor. Clopidogrel is used to inhibit blood clots in coronary artery disease, peripheral vascular disease, and cerebrovascular disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.