Learn More
Apexbio Technology LLC Clopidogrel 120202-66-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A51835.1
Encompass_Preferred
Clopidogrel (CAS 120202-66-6) is a selective antagonist targeting the platelet P2Y receptor thereby preventing ADP-mediated platelet activation and aggregation Through this mechanism clopidogrel exhibits pronounced antithrombotic properties reducing thrombus formation It is the pharmacologically active enantiomer derived from racemic ( )-clopidogrel hydrochloride Clopidogrel is widely employed in biomedical research examining platelet function thrombosis pathways and therapeutic interventions targeting cardiovascular disease
Catalog No. 50-000-61619