Learn More
Med Vet International Clopidogrel Bisulfate Tablets (Plavix), 75mg, 90ct
Catalog No. 502828020
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Clopidogrel bisulfate is a thienopyridine class of drug that inhibits platelet aggregation and thus inhibits aspects of blood clotting used to treat patients with acute coronary syndrome, myocardial infarction (MI), peripheral vascular disease and some stroke (ischemic type) patients.
Catalog No. 50-282-8020 Supplier Med Vet International Supplier No. RX50228012490
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Clopidogrel bisulfate is a thienopyridine class of drug that inhibits platelet aggregation and thus inhibits aspects of blood clotting used to treat patients with acute coronary syndrome, myocardial infarction (MI), peripheral vascular disease and some stroke (ischemic type) patients.