Learn More
Medchemexpress LLC HY-17459 100mg , Clopidogrel (hydrogen sulfate) CAS:120202-66-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY17459100MG
Encompass_Preferred
HY-17459 100mg Clopidogrel (hydrogen sulfate) CAS: 120202-66-6 Clopidogrel hydrogen sulfate is an antiplatelet agent to prevent blood clots. Clopidogrel hydrogen sulfate inhibits CYP2B6 and CYP2C19 with IC50s of 18.2 nM and 524 nM, respectively. Clopidogrel hydrogen sulfate is a potent antithrombotic agent that inhibits ADP-induced platelet aggregation.Clopidogrel hydrogen sulfate also is an orally active P2Y(12) inhibitor[5]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1897