Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clopidogrel-related Compound B (hydrochloride) | 144750-52-7 | 99.7% | 358.28 | 50 MG
SDP

Catalog No. 5000452191
Encompass_Preferred

Clopidogrel-related Compound B hydrochloride is an impurity of Clopidogrel. It is a solid with a white to yellow appearance, typically used for laboratory research.

  • High purity at 99.67%
  • Molecular weight of 358.28
  • Chemical formula is C16H17Cl2NO2S
  • Identified by CAS number 144750-52-7

Catalog No. 50-004-52191 Supplier Medchemexpress LLC Supplier No. HYW01111450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clopidogrel-related Compound B hydrochloride is an impurity of Clopidogrel. It is a solid with a white to yellow appearance, typically used for laboratory research.

  • High purity at 99.67%
  • Molecular weight of 358.28
  • Chemical formula is C16H17Cl2NO2S
  • Identified by CAS number 144750-52-7

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.