Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clopidogrel Related Compound C Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111018068 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
40 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8068 40 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8068 Supplier MilliporeSigma Supelco Supplier No. PHR199240MG
Add to Cart
Add to Cart

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

TRUSTED_SUSTAINABILITY

Specifications

CAS 120202-71-3
Linear Formula C16H16 ClNO2 S · H2 SO4
MDL Number MFCD17215518
Synonym (R)-Clopidogrel bisulfate; (R)-Methyl (2-chlorophenyl)(6,7-dihydro-4 H-thieno[3,2-c]pyridin-5-yl)acetate hydrogensulfate
Type Certified Reference Material
Quantity 40 mg
Formula Weight 419.90
Grade Certified Reference Material
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.