Learn More
Cayman Chemical Clopidogrel-related Compound B (hydrochloride), Purity: ≥98%, CAS: 144750-52-7, 5 mg
SDP
Catalog No. 502912194
Encompass_Preferred
A potential impurity in commercial preparations of (S)-(+)-clopidogrel
Catalog No. 50-291-2194 Supplier Cayman Chemical Supplier No. 365075 MG
A potential impurity in commercial preparations of (S)-(+)-clopidogrel