Learn More
Cayman Chemical -Cloprostenol 10mg
SDP
Catalog No. 502966693
Encompass_Preferred
A synthetic analog of PGF2α; an FP receptor agonist and a potent luteolytic agent in rats and hamsters; the optically active, 15(R) enantiomer of cloprostenol responsible for the majority of its biological activity
Catalog No. 50-296-6693 Supplier Cayman Chemical Supplier No. 1676510 MG
A synthetic analog of PGF2α; an FP receptor agonist and a potent luteolytic agent in rats and hamsters; the optically active, 15(R) enantiomer of cloprostenol responsible for the majority of its biological activity