Learn More
Biomatik Corporation Cloprostenol (CP) ELISA Kit
Supplier:  Biomatik Corporation EKU10720
Encompass_Preferred
Cloprostenol (CP) ELISA Kit for detecting CP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Universal. Detection Range: 1.23-100ng/ml. Sensitivity: 0.52ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8304