Learn More
Medchemexpress LLC HY-108415 10mg Medchemexpress, Cloprostenol sodium salt CAS:55028-72-3 Purity:>98%
SDP
Catalog No. 501968543
Encompass_Preferred
Medchemexpress, HY-108415 10mg Cloprostenol sodium salt CAS:55028-72-3 Cloprostenol sodium salt (ICI 80996 sodium salt) is a potent synthetic prostaglandin analogue, acts as a luteolytic agent, and is a PGF2α receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8543 Supplier Medchemexpress LLC Supplier No. HY10841510MG
Medchemexpress, HY-108415 10mg Cloprostenol sodium salt CAS:55028-72-3 Cloprostenol sodium salt (ICI 80996 sodium salt) is a potent synthetic prostaglandin analogue, acts as a luteolytic agent, and is a PGF2α receptor agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.