Learn More
Medchemexpress LLC Cloprostenol sodium salt | 55028-72-3 | 99.5% | 446.90 | 10 MM 1 ML
SDP
Catalog No. 5000346848
Encompass_Preferred
Cloprostenol sodium salt (ICI 80996 sodium salt) is a potent synthetic prostaglandin analogue that acts as a luteolytic agent and is a PGF2α receptor agonist. It is intended for research use only.
  • Potent synthetic prostaglandin analogue.
  • Acts as a luteolytic agent.
  • PGF2α receptor agonist.
  • In vitro, cloprostenol (0.1 μM) counteracts the adipogenic effects of statil, on both intracellular lipid accumulation and expression of transcripts for proadipogenic factors C/EBPα and PPARγ after treatment for 6 days.
  • In vivo, cloprostenol sodium salt (25 μg) decreases plasma progesterone in pregnant rats.
Catalog No. 50-003-46848 Supplier Medchemexpress LLC Supplier No. HY10841510MM/1ML
Cloprostenol sodium salt (ICI 80996 sodium salt) is a potent synthetic prostaglandin analogue that acts as a luteolytic agent and is a PGF2α receptor agonist. It is intended for research use only.
  • Potent synthetic prostaglandin analogue.
  • Acts as a luteolytic agent.
  • PGF2α receptor agonist.
  • In vitro, cloprostenol (0.1 μM) counteracts the adipogenic effects of statil, on both intracellular lipid accumulation and expression of transcripts for proadipogenic factors C/EBPα and PPARγ after treatment for 6 days.
  • In vivo, cloprostenol sodium salt (25 μg) decreases plasma progesterone in pregnant rats.