Learn More
TARGETMOL CHEMICALS INC Cloprostenol sodium salt 5MG
SDP
Catalog No. 502880113
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cloprostenol sodium salt (ICI 80996 sodium salt) is a more water-soluble, crystalline form of cloprostenol compared to the free acid and is a synthetic analog of prostaglandin F2alpha. Purity 98%
Catalog No. 50-288-0113 Supplier TARGETMOL CHEMICALS INC Supplier No. T58475MG
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Cloprostenol sodium salt (ICI 80996 sodium salt) is a more water-soluble, crystalline form of cloprostenol compared to the free acid and is a synthetic analog of prostaglandin F2alpha. Purity 98%