Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clopyralid | 1702-17-6 | MFCD00078655 | 97.0% | 192.00 g·mol-1 | C6H3Cl2NO2 | 250 MG
SDP

Catalog No. 5000303646
Encompass_Preferred

Clopyralid (Standard) is an analytical reference standard of the herbicide clopyralid (CAS 1702-17-6). It is supplied as a 250 mg vial with 97.0% purity for laboratory analytical and method development use (HPLC, GC, MS). For research use only; not for human use or clinical applications.

  • Analytical standard for clopyralid (CAS 1702-17-6).
  • High purity: 97.0%.
  • Suitable for HPLC, GC, and MS method development and validation.
  • Molecular formula C6H3Cl2NO2; molecular weight 192.00 g·mol-1.
  • Supplied as a 250 mg packaging for analytical testing.
  • Intended for research use only; not for human use.
  • Storage: follow the certificate of analysis; room temperature shipping noted.

Catalog No. 50-003-03646 Supplier Medchemexpress LLC Supplier No. 250MGHYW018749R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clopyralid (Standard) is an analytical reference standard of the herbicide clopyralid (CAS 1702-17-6). It is supplied as a 250 mg vial with 97.0% purity for laboratory analytical and method development use (HPLC, GC, MS). For research use only; not for human use or clinical applications.

  • Analytical standard for clopyralid (CAS 1702-17-6).
  • High purity: 97.0%.
  • Suitable for HPLC, GC, and MS method development and validation.
  • Molecular formula C6H3Cl2NO2; molecular weight 192.00 g·mol-1.
  • Supplied as a 250 mg packaging for analytical testing.
  • Intended for research use only; not for human use.
  • Storage: follow the certificate of analysis; room temperature shipping noted.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.