Learn More
Dexsil Corporation CLOR-D-TECT Q4000 80/CS
Catalog No. NC0311311
Encompass
Field Test Kit for Quantifying Total (Halogens) Chlorine in Used Oil between 200-4000ppm
Catalog No. NC0311311 Supplier Dexsil Corporation Supplier No. Q4000CS
Field Test Kit for Quantifying Total (Halogens) Chlorine in Used Oil between 200-4000ppm