Learn More
Dexsil Corporation Clor-N-Oil 50 ppm, 20 kits
Catalog No. 50465276
Encompass
Designed to be used in the field, Clor-N-Oil is a fast, accurate method to test electrical insulating fluids for the presence of PCB. Each pocket-sized kit contains everything necessary to perform the test in less than 5 minutes. All premeasured reagents are sealed in glass ampules for safe, fast, consistent and accurate results. Screening transformer oil with Clor-N-Oil can eliminate non-PCB fluids from costly laboratory analysis and allows a greater number of samples to be tested at less cost. 20kits
Catalog No. 50-465-276 Supplier Dexsil Corporation Supplier No. CL050SP
Designed to be used in the field, Clor-N-Oil is a fast, accurate method to test electrical insulating fluids for the presence of PCB. Each pocket-sized kit contains everything necessary to perform the test in less than 5 minutes. All premeasured reagents are sealed in glass ampules for safe, fast, consistent and accurate results. Screening transformer oil with Clor-N-Oil can eliminate non-PCB fluids from costly laboratory analysis and allows a greater number of samples to be tested at less cost. 20kits