Learn More
Cayman Chemical Chlorcyclizine (hydrochloride), Purity: ≥98%, CAS: 14362-31-3, 100 mg
SDP
Catalog No. 502982730
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 9 nM) that is effective against HCV (EC50 = 44 nM), preventing viral entry into host cells
Catalog No. 50-298-2730 Supplier Cayman Chemical Supplier No. 19239100 MG
A histamine H1 receptor antagonist (Ki = 9 nM) that is effective against HCV (EC50 = 44 nM), preventing viral entry into host cells