Learn More
Medchemexpress LLC HY-14197A 10mg Medchemexpress, Clorgyline hydrochloride CAS:17780-75-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY14197A10MG
Encompass_Preferred
Medchemexpress, HY-14197A 10mg Clorgyline hydrochloride CAS:17780-75-5 Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) that is used in scientific research; structurally related to Pargyline. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0518