Learn More
Medchemexpress LLC Clorgyline hydrochloride | 17780-75-5 | 99.9% | C13H16Cl3NO | 1 ML
SDP
Catalog No. 5000356801
Encompass_Preferred
Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) that is used in scientific research. It exhibits minimal effect on the amounts of conjugated dopamine (DA) present in superfusate of slices from rat striatum. This compound contains an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Irreversible and selective monoamine oxidase A (MAO-A) inhibitor
  • Used for scientific research
  • Minimal impact on conjugated dopamine (DA) in rat striatum slices
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Stable under recommended storage conditions
Catalog No. 50-003-56801 Supplier Medchemexpress LLC Supplier No. HY14197A10MM/1ML
Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) that is used in scientific research. It exhibits minimal effect on the amounts of conjugated dopamine (DA) present in superfusate of slices from rat striatum. This compound contains an alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups.
  • Irreversible and selective monoamine oxidase A (MAO-A) inhibitor
  • Used for scientific research
  • Minimal impact on conjugated dopamine (DA) in rat striatum slices
  • Contains an alkyne group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
  • Stable under recommended storage conditions