Learn More
Medchemexpress LLC Clorgyline hydrochloride | 17780-75-5 | 99.9% | C13H16Cl3NO | 100 MG
SDP
Catalog No. 5000353257
Encompass_Preferred
Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) used in scientific research. It is structurally related to Pargyline. It has little effect on the amounts of conjugated dopamine (DA) in superfusate of slices from rat striatum. It contains an Alkyne group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. This product is for research use only, not sold to patients.
  • Irreversible and selective inhibitor of monoamine oxidase A (MAO-A)
  • Used in scientific research
  • Structurally related to Pargyline
  • Has little effect on conjugated dopamine (DA) in superfusate of slices from rat striatum
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups
Catalog No. 50-003-53257 Supplier Medchemexpress LLC Supplier No. HY14197A100MG
Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) used in scientific research. It is structurally related to Pargyline. It has little effect on the amounts of conjugated dopamine (DA) in superfusate of slices from rat striatum. It contains an Alkyne group, allowing it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. This product is for research use only, not sold to patients.
  • Irreversible and selective inhibitor of monoamine oxidase A (MAO-A)
  • Used in scientific research
  • Structurally related to Pargyline
  • Has little effect on conjugated dopamine (DA) in superfusate of slices from rat striatum
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups