Learn More
Medchemexpress LLC Clorgyline hydrochloride | 17780-75-5 | 99.9% | C13H16Cl3NO | 50 MG
SDP
Catalog No. 5000353026
Encompass_Preferred
Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) used in scientific research. It has minimal effect on conjugated dopamine (DA) amounts in superfusates of rat striatum slices. This compound contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Irreversible and selective inhibitor of monoamine oxidase A (MAO-A)
  • Structurally related to Pargyline
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups
  • Increases nicotine infusions and motivation to obtain nicotine in rats at 2 mg/kg daily for 28 days
  • Pretreatment with 1 mg/kg intravenously for 1 hour has little effect on the efflux of conjugated dopamine (DA) and p-tyramine-evoked release of conjugated DA from slices of rat striatum
Catalog No. 50-003-53026 Supplier Medchemexpress LLC Supplier No. HY14197A50MG
Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) used in scientific research. It has minimal effect on conjugated dopamine (DA) amounts in superfusates of rat striatum slices. This compound contains an Alkyne group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Irreversible and selective inhibitor of monoamine oxidase A (MAO-A)
  • Structurally related to Pargyline
  • Contains an Alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Azide groups
  • Increases nicotine infusions and motivation to obtain nicotine in rats at 2 mg/kg daily for 28 days
  • Pretreatment with 1 mg/kg intravenously for 1 hour has little effect on the efflux of conjugated dopamine (DA) and p-tyramine-evoked release of conjugated DA from slices of rat striatum