Learn More
Medchemexpress LLC Clorgyline hydrochloride | 17780-75-5 | MFCD00052012 | 99.9% | 308.63 g/mol | C13H16Cl3NO | 5 MG
SDP
Catalog No. 5000221598
Encompass_Preferred
Clorgyline hydrochloride is an irreversible, selective monoamine oxidase A (MAO-A) inhibitor used in biochemical and pharmacological research. Supplied as the hydrochloride salt with high reported purity, it is used to probe MAO-A activity, neurotransmitter metabolism, and related signaling pathways. The molecule's alkyne functionality also enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) applications.
  • Selective irreversible MAO-A inhibitor.
  • High purity suitable for research applications.
  • Available in small, precise quantities for laboratory use.
  • Contains an alkyne group compatible with CuAAC click chemistry.
  • Useful for studies of neurotransmitter metabolism and MAO-A function.
Catalog No. 50-002-21598 Supplier Medchemexpress LLC Supplier No. HY14197A5MG
Clorgyline hydrochloride is an irreversible, selective monoamine oxidase A (MAO-A) inhibitor used in biochemical and pharmacological research. Supplied as the hydrochloride salt with high reported purity, it is used to probe MAO-A activity, neurotransmitter metabolism, and related signaling pathways. The molecule's alkyne functionality also enables copper-catalyzed azide-alkyne cycloaddition (CuAAC) applications.
  • Selective irreversible MAO-A inhibitor.
  • High purity suitable for research applications.
  • Available in small, precise quantities for laboratory use.
  • Contains an alkyne group compatible with CuAAC click chemistry.
  • Useful for studies of neurotransmitter metabolism and MAO-A function.