Learn More
Medchemexpress LLC Clorgyline hydrochloride | 17780-75-5 | MFCD00052012 | 99.9% | C13H16Cl3NO | 500 MG
SDP
Catalog No. 5000354972
Encompass_Preferred
Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) used in scientific research. It is structurally related to Pargyline. This compound has little effect on the amounts of conjugated dopamine (DA) present in superfusate of slices from rat striatum. Clorgyline hydrochloride contains an alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This product is for research use only and not sold to patients.
  • Irreversible and selective inhibitor of monoamine oxidase A (MAO-A)
  • Increases nicotine infusions and motivation to obtain nicotine in rats
  • Has little effect on the efflux of conjugated dopamine (DA) and p-tyramine-evoked release of conjugated DA from slices of rat striatum
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)
Catalog No. 50-003-54972 Supplier Medchemexpress LLC Supplier No. HY14197A500MG
Clorgyline hydrochloride is an irreversible and selective inhibitor of monoamine oxidase A (MAO-A) used in scientific research. It is structurally related to Pargyline. This compound has little effect on the amounts of conjugated dopamine (DA) present in superfusate of slices from rat striatum. Clorgyline hydrochloride contains an alkyne group and can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing azide groups. This product is for research use only and not sold to patients.
  • Irreversible and selective inhibitor of monoamine oxidase A (MAO-A)
  • Increases nicotine infusions and motivation to obtain nicotine in rats
  • Has little effect on the efflux of conjugated dopamine (DA) and p-tyramine-evoked release of conjugated DA from slices of rat striatum
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc)