Learn More
Abcam Clorgyline hydrochloride, MAO-A inhibitor, 10MG
Supplier:  Abcam AB14564610MG
Encompass_Preferred
MW 308.6 Da, Purity >98%. Selective, irreversible MAO-A inhibitor (Ki values are 0.054 and 58 μM for MAO-A and MAO-B respectively). Potent σ1 receptor inhibitor (IC₅₀ = 31 nM). Lysine-specific demethylase 1 inhibitor. Centrally active in vivo. .
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1345572