Learn More
Cayman Chemical Clorgyline (hydrochloride), Purity: ≥98%, CAS: 17780-75-5, 10 mg
SDP
Catalog No. 502971764
Encompass_Preferred
A potent MAO inhibitor that preferentially targets MAO-A over MAO-B (Kis = 0.054 and 58 µM, respectively); also inhibits the sigma receptor σ1 on Jurkat human T lymphocyte cells (IC50 = 31 nM)
Catalog No. 50-297-1764 Supplier Cayman Chemical Supplier No. 1592510 MG
A potent MAO inhibitor that preferentially targets MAO-A over MAO-B (Kis = 0.054 and 58 µM, respectively); also inhibits the sigma receptor σ1 on Jurkat human T lymphocyte cells (IC50 = 31 nM)