Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cloricromen hydrochl 100mg | 74697-28-2 | 100 MG
SDP

Catalog No. 5000308235
Encompass_Preferred

Cloricromen hydrochloride is the hydrochloride salt of cloricromen, a platelet aggregation inhibitor used in research to inhibit platelet aggregation in humans and in experimental thrombosis.

  • Platelet aggregation inhibitor used in thrombosis research.
  • Reported purity 99.82%.
  • Molecular formula C20H27Cl2NO5; molecular weight 432.34.
  • White to off-white solid appearance.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple quantities including 100 MG for dosing flexibility.

Catalog No. 50-003-08235 Supplier Medchemexpress LLC Supplier No. HY116678A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cloricromen hydrochloride is the hydrochloride salt of cloricromen, a platelet aggregation inhibitor used in research to inhibit platelet aggregation in humans and in experimental thrombosis.

  • Platelet aggregation inhibitor used in thrombosis research.
  • Reported purity 99.82%.
  • Molecular formula C20H27Cl2NO5; molecular weight 432.34.
  • White to off-white solid appearance.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Available in multiple quantities including 100 MG for dosing flexibility.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.