Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cloricromen hydrochl 10mM 1mL | 74697-28-2 | 1 ML
SDP

Catalog No. 5000308148
Encompass_Preferred

Cloricromen hydrochloride is a small-molecule platelet aggregation inhibitor supplied as a solid or as a ready-to-use solution in DMSO. It is used in in vitro platelet aggregation assays and experimental thrombosis models for research applications.

  • Platelet aggregation inhibitor for research use.
  • Available as solid and as 10 mM, 1 mL solution in DMSO.
  • High reported purity (99.8%).
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic treatment.
  • Store powder at 4 °C (up to 2 years) or -20 °C (up to 3 years); store solutions at -20 °C (1 month) or -80 °C (6 months).
  • Molecular weight 432.34 g/mol; formula C20H27Cl2NO5.

Catalog No. 50-003-08148 Supplier Medchemexpress LLC Supplier No. HY116678A10MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cloricromen hydrochloride is a small-molecule platelet aggregation inhibitor supplied as a solid or as a ready-to-use solution in DMSO. It is used in in vitro platelet aggregation assays and experimental thrombosis models for research applications.

  • Platelet aggregation inhibitor for research use.
  • Available as solid and as 10 mM, 1 mL solution in DMSO.
  • High reported purity (99.8%).
  • Soluble in DMSO at 100 mg/mL; may require ultrasonic treatment.
  • Store powder at 4 °C (up to 2 years) or -20 °C (up to 3 years); store solutions at -20 °C (1 month) or -80 °C (6 months).
  • Molecular weight 432.34 g/mol; formula C20H27Cl2NO5.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.