Learn More
Medchemexpress LLC Cloricromen hydrochl 25mg | 74697-28-2 | 432.34 g/mol | C20H26ClNO5·HCl | 25 MG
SDP
Catalog No. 5000308177
Encompass_Preferred
Cloricromen hydrochloride is the hydrochloride salt of cloricromen, a research compound that acts as a platelet aggregation inhibitor used in studies of platelet function and experimental thrombosis.
  • High purity (99.82%).
  • White solid hydrochloride salt.
  • Acts as a platelet aggregation inhibitor for studies of platelet function and experimental thrombosis.
  • Suitable for biochemical and pharmacological research.
  • Identified by CAS 74697-28-2.
Catalog No. 50-003-08177 Supplier Medchemexpress LLC Supplier No. HY116678A25MG
Cloricromen hydrochloride is the hydrochloride salt of cloricromen, a research compound that acts as a platelet aggregation inhibitor used in studies of platelet function and experimental thrombosis.
  • High purity (99.82%).
  • White solid hydrochloride salt.
  • Acts as a platelet aggregation inhibitor for studies of platelet function and experimental thrombosis.
  • Suitable for biochemical and pharmacological research.
  • Identified by CAS 74697-28-2.