Learn More
Cayman Chemical CLOrmethIazolehydroCLOrId 10mg
SDP
Catalog No. 502947672
Encompass_Preferred
A potentiator of GABAA receptors; potentiates GABA-induced currents in L-M(TK-) cells expressing α1β1γ2 or α1β2γ2 subunit-containing GABAA receptors (EC50s = 0.8 and 3.4 µM, respectively); protects against MES-induced seizures in mice (ED50 = 130.8 mg/kg); reduces infarct volume in a permanent rat model of MCAO-induced focal ischemia
Catalog No. 50-294-7672 Supplier Cayman Chemical Supplier No. 3343710 MG
A potentiator of GABAA receptors; potentiates GABA-induced currents in L-M(TK-) cells expressing α1β1γ2 or α1β2γ2 subunit-containing GABAA receptors (EC50s = 0.8 and 3.4 µM, respectively); protects against MES-induced seizures in mice (ED50 = 130.8 mg/kg); reduces infarct volume in a permanent rat model of MCAO-induced focal ischemia