Learn More
TARGETMOL CHEMICALS INC Cloroperone hydrochloride 1MG
SDP
Catalog No. 502876777
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Cloroperone hydrochloride (AHR-6134) has anxiolytic activity and may be used in the study of neurologic disorders. Purity 98.56%
Catalog No. 50-287-6777 Supplier TARGETMOL CHEMICALS INC Supplier No. T309841MG
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Cloroperone hydrochloride (AHR-6134) has anxiolytic activity and may be used in the study of neurologic disorders. Purity 98.56%