Learn More
Cayman Chemical CLOrophenol Red DGalactop 50mg
SDP
Catalog No. 502990476
Encompass_Preferred
A colorimetric β-galactosidase substrate; upon enzymatic cleavage by β-galactosidase, chlorophenol red is released, which can be quantified by colorimetric detection at 570 nm as a measure of β-galactosidase activity
Catalog No. 50-299-0476 Supplier Cayman Chemical Supplier No. 2970750 MG
A colorimetric β-galactosidase substrate; upon enzymatic cleavage by β-galactosidase, chlorophenol red is released, which can be quantified by colorimetric detection at 570 nm as a measure of β-galactosidase activity