Learn More
Essendant Bleach Germicidal Cleaner, 128 oz Refill Bottle, 4/Carton
Catalog No. NC0034214
Encompass_Preferred
Healthcare professionals trust Clorox Healthcare Bleach Germicidal Cleaner to help them overcome infection control challenges in a constantly changing healthcare environment. Clorox Healthcare Bleach Germicidal Cleaner is a ready to use, 1 step cleaner- disinfectant containing a 1:10 bleach dilution to meet CDC and OSHA guidelines for disinfecting surfaces in healthcare settings. This cleaner disinfectant is EPA registered to kill a total of 50 microorganisms, including HAI causing bacteria like MRSA and VRE, outbreak causing viruses such as Influenza A and B viruses, Measles virus and Norovirus and fungi in 1 minute. This disinfectant kills 99.9999% of C. difficile spores in 3 minutes, according to the current EPA recommended testing standard (June 2014). The ready-to-use disinfectant eliminates compliance risks and labor costs associated with dilutable disinfectants. Designed for easy refilling to meet needs of heavy users.
Catalog No. NC0034214 Supplier Essendant Supplier No. CLO68978
Healthcare professionals trust Clorox Healthcare Bleach Germicidal Cleaner to help them overcome infection control challenges in a constantly changing healthcare environment. Clorox Healthcare Bleach Germicidal Cleaner is a ready to use, 1 step cleaner- disinfectant containing a 1:10 bleach dilution to meet CDC and OSHA guidelines for disinfecting surfaces in healthcare settings. This cleaner disinfectant is EPA registered to kill a total of 50 microorganisms, including HAI causing bacteria like MRSA and VRE, outbreak causing viruses such as Influenza A and B viruses, Measles virus and Norovirus and fungi in 1 minute. This disinfectant kills 99.9999% of C. difficile spores in 3 minutes, according to the current EPA recommended testing standard (June 2014). The ready-to-use disinfectant eliminates compliance risks and labor costs associated with dilutable disinfectants. Designed for easy refilling to meet needs of heavy users.