Learn More
Enterprise Technology Solutions Clorox Company Cleaner,Multi-surface,Lavender,Conc,80oz,PE
SDP
Catalog No. 5000832271
Encompass_Preferred
With Pine-Sol Multi-Surface Cleaner from CloroxPro, you only need one product to fight tough dirt, grease and grime. Twice-concentrated formula delivers more cleaning power per drop than previous Pine-Sol Pine formula when used as directed on hard, nonporous surfaces. Easier to handle and easier to store, the space-saving bottle gives you more cleaning solution in a smaller bottle made of 30 percent less plastic vs, previous Pine-Sol 144 oz. bottle. Multipurpose cleaner cleans, deodorizes and disinfects in one step, killing 99.9 percent of germs (salmonella, Staph, Influenza A virus and SARS-CoV-2 ) and leaving a classic pine scent. It cuts through grime and soap scum and is safe for use on wood, tile, stone, glass and more when used as directed on hard, nonporous surfaces. Do not use on marble, aluminum, copper or wood that is unfinished, unsealed, unpainted, waxed, oiled or worn.
Catalog No. 50-008-32271 Supplier Enterprise Technology Solutions Supplier No. CLO60608
With Pine-Sol Multi-Surface Cleaner from CloroxPro, you only need one product to fight tough dirt, grease and grime. Twice-concentrated formula delivers more cleaning power per drop than previous Pine-Sol Pine formula when used as directed on hard, nonporous surfaces. Easier to handle and easier to store, the space-saving bottle gives you more cleaning solution in a smaller bottle made of 30 percent less plastic vs, previous Pine-Sol 144 oz. bottle. Multipurpose cleaner cleans, deodorizes and disinfects in one step, killing 99.9 percent of germs (salmonella, Staph, Influenza A virus and SARS-CoV-2 ) and leaving a classic pine scent. It cuts through grime and soap scum and is safe for use on wood, tile, stone, glass and more when used as directed on hard, nonporous surfaces. Do not use on marble, aluminum, copper or wood that is unfinished, unsealed, unpainted, waxed, oiled or worn.