Learn More
Enterprise Technology Solutions Clorox Company Cleaner,Multisurface,Liquid,Conc,Lemon Fresh,40oz,YW
SDP
Catalog No. 5000833076
Encompass_Preferred
With Pine-Sol Multi-Surface Cleaner, you only need one cleaner to fight tough dirt, grease and grime. This twice-concentrated formula delivers more cleaning power per drop and cleans five times better than the previous Pine-Sol Scented Formula This multipurpose cleaner cleans right the first time, removing stains and deodorizing soft surfaces while leaving a lemon fresh scent. This powerful degreaser cuts through grime and soap scum fast and is safe for use on wood, tile, stone, porcelain and glass and more when used as directed on hard, nonporous surfaces. Do not use on marble, aluminum, copper or wood that is unfinished, unsealed, unpainted, waxed, oiled or worn.
Catalog No. 50-008-33076 Supplier Enterprise Technology Solutions Supplier No. CLO60165
With Pine-Sol Multi-Surface Cleaner, you only need one cleaner to fight tough dirt, grease and grime. This twice-concentrated formula delivers more cleaning power per drop and cleans five times better than the previous Pine-Sol Scented Formula This multipurpose cleaner cleans right the first time, removing stains and deodorizing soft surfaces while leaving a lemon fresh scent. This powerful degreaser cuts through grime and soap scum fast and is safe for use on wood, tile, stone, porcelain and glass and more when used as directed on hard, nonporous surfaces. Do not use on marble, aluminum, copper or wood that is unfinished, unsealed, unpainted, waxed, oiled or worn.