Learn More
Enterprise Technology Solutions Clorox Company Formula 409 Multisurface Cleaner, Original Scent, 64 fl oz, White
SDP
Catalog No. 5000832652
Encompass_Preferred
Formula 409 Multisurface Cleaner is the ultimate cleaner, degreaser and disinfectant for your home. Antibacterial formula quickly cuts through grease and grime and disinfects as you wipe, killing more than 99.9 percent of germs on hard, nonporous surfaces, including COVID-19 Virus, salmonella, E. coli, Staph, Influenza A Virus and Herpes Simplex Virus Type 2. When used as directed, this one cleaner works on many surfaces: stainless steel, synthetic marble, glazed ceramic tile, linoleum, countertops, stovetops, sinks, floors, cabinets, tubs and walls. A potable rinse is required for food-contact surfaces. No-bleach formula deodorizes as you clean, leaving a fresh scent. It works great both indoors and outdoors on a wide range of hard, nonporous surfaces. One 64 oz. refill can fill two 32 oz. spray bottles.
Catalog No. 50-008-32652 Supplier Enterprise Technology Solutions Supplier No. CLO00636
Formula 409 Multisurface Cleaner is the ultimate cleaner, degreaser and disinfectant for your home. Antibacterial formula quickly cuts through grease and grime and disinfects as you wipe, killing more than 99.9 percent of germs on hard, nonporous surfaces, including COVID-19 Virus, salmonella, E. coli, Staph, Influenza A Virus and Herpes Simplex Virus Type 2. When used as directed, this one cleaner works on many surfaces: stainless steel, synthetic marble, glazed ceramic tile, linoleum, countertops, stovetops, sinks, floors, cabinets, tubs and walls. A potable rinse is required for food-contact surfaces. No-bleach formula deodorizes as you clean, leaving a fresh scent. It works great both indoors and outdoors on a wide range of hard, nonporous surfaces. One 64 oz. refill can fill two 32 oz. spray bottles.