Learn More
Enterprise Technology Solutions Clorox Company Hand Sanitizer, Bleach-free, 16.9 oz Pump Bottle, 864/PL, CL
SDP
Catalog No. 5000790437
Encompass_Preferred
Clorox Hand Sanitizer is formulated with 71 percent ethyl alcohol and provides broad-spectrum antimicrobial activity, killing greater than 99.999 percent of germs in as little as 15 seconds. It contains premium emollients to keep hands soft instead of leaving them sticky or greasy like gels and foams can. From Clorox Commercial Solutions, this sanitizer spray is perfect for use in office breakrooms, lobbies, classrooms, restaurants and healthcare facilities.
Catalog No. 50-007-90437 Supplier Enterprise Technology Solutions Supplier No. CLO02176PL
Clorox Hand Sanitizer is formulated with 71 percent ethyl alcohol and provides broad-spectrum antimicrobial activity, killing greater than 99.999 percent of germs in as little as 15 seconds. It contains premium emollients to keep hands soft instead of leaving them sticky or greasy like gels and foams can. From Clorox Commercial Solutions, this sanitizer spray is perfect for use in office breakrooms, lobbies, classrooms, restaurants and healthcare facilities.